Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023876.3 | NP_076365.2 | 4 | 408 | ||
GenBank mRNA | AJ276004.1 | - | 4 | 407 | ||
AK020742.1 | - | 4 | 416 | |||
AK034040.1 | - | 4 | 413 | |||
AK034529.1 | - | 4 | 414 | |||
AK037104.1 | - | 4 | 432 | |||
AK039447.1 | - | 4 | 420 | |||
AK039758.1 | - | 4 | 409 | |||
AK079069.1 | - | 4 | 423 | |||
AK168964.1 | - | 4 | 472 | |||
EF410086.1 | - | 272 |