Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLR9 SIRNA Assays ›
Gene Symbol:
Tlr9
Gene Name:
toll-like receptor 9
Gene Aliases:
-
Chromosome Location:
Chr.9: 106222598 - 106226876 on Build GRCm38
UniGene ID:
Mm.44889
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s211982
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tlr9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031178.2
NP_112455.2
2
2469
GenBank mRNA
AB045181.1
-
2
2469
AF314224.1
-
2
2448
AF348140.1
-
2
2402
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microglial cell activation
regulation of protein phosphorylation
toll-like receptor signaling pathway
response to molecule of bacterial origin
immune system process
regulation of dendritic cell cytokine production
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
immune response
signal transduction
I-kappaB phosphorylation
response to virus
positive regulation of autophagy
positive regulation of gene expression
maintenance of gastrointestinal epithelium
negative regulation of NF-kappaB transcription factor activity
tumor necrosis factor production
negative regulation of interleukin-6 production
negative regulation of interleukin-8 production
positive regulation of chemokine production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-beta production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-18 production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
negative regulation of toll-like receptor signaling pathway
positive regulation of toll-like receptor signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of NF-kappaB import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
positive regulation of interferon-gamma biosynthetic process
innate immune response
positive regulation of interferon-alpha biosynthetic process
positive regulation of interferon-beta biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
regulation of cytokine secretion
regulation of inflammatory response
positive regulation of inflammatory response
defense response to Gram-negative bacterium
regulation of B cell activation
positive regulation of NF-kappaB transcription factor activity
defense response to virus
positive regulation of nitric-oxide synthase biosynthetic process
negative regulation of calcium-transporting ATPase activity
transmembrane signaling receptor activity
interleukin-1 receptor binding
protein binding
siRNA binding
unmethylated CpG binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0