Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEPT9 SIRNA Assays ›
          Gene Symbol:
          SEPT9
          Gene Name:
          septin 9
          Gene Aliases:
          AF17q25, MSF, MSF1, NAPB, PNUTL4, SINT1, SeptD1
          Chromosome Location:
          Chr.17: 77281410 - 77500596 on Build GRCh38
          UniGene ID:
          Hs.440932
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s21222

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEPT9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113491.1 NP_001106963.1 5, 6 1171
          NM_001113492.1 NP_001106964.1 5, 6 1346
          NM_001113493.1 NP_001106965.1 4, 5 1332
          NM_001113494.1 NP_001106966.1 4, 5 1285
          NM_001113495.1 NP_001106967.1 3, 4 985
          NM_001113496.1 NP_001106968.1 3, 4 580
          NM_001293695.1 NP_001280624.1 4, 5 1114
          NM_001293696.1 NP_001280625.1 3, 4 396
          NM_001293697.1 NP_001280626.1 3, 4 609
          NM_001293698.1 NP_001280627.1 3, 4 695
          NM_006640.4 NP_006631.2 4, 5 1803
          XM_005256962.1 XP_005257019.1 3, 4 453
          XM_006721643.2 XP_006721706.1 6, 7 1274
          XM_006721644.1 XP_006721707.1 3, 4 496
          XM_011524204.1 XP_011522506.1 6, 7 1371
          XM_011524206.1 XP_011522508.1 4, 5 1091
          XM_011524207.1 XP_011522509.1 4, 5 1104
          XM_011524208.2 XP_011522510.1 3, 4 362
          XM_017024031.1 XP_016879520.1 4, 5 703
          XM_017024032.1 XP_016879521.1 3, 4 702
          GenBank mRNA AB023208.1 - 4, 5 1285
          AF123052.1 - 4, 5 1766
          AF142408.1 - 4, 5 1248
          AF189712.2 - 6, 7 1298
          AF189713.3 - 5, 6 1141
          AJ312319.1 - 4, 5 1332
          AJ312320.1 - 3, 4 374
          AJ312321.1 - 4, 5 1803
          AJ312322.1 - 4, 5 1274
          AK022493.1 - 3, 4 362
          AK056495.1 - 3, 4 985
          AK290368.1 - 5, 6 1346
          AK299828.1 - 3, 4 609
          AK300268.1 - 6, 7 1005
          AK300270.1 - 4, 5 1073
          AK303449.1 - 3, 4 389
          AK304143.1 - 4, 5 1186
          AK316473.1 - 3, 4 695
          BC021192.2 - 4, 5 1026 3346534
          BC054004.1 - 6, 7 1001 6146448
          BT007215.1 - 4, 5 991
          CN393537.1 - 3, 4 388
          DC410238.1 - 3, 4 397
          KU178538.1 - 3, 4 346
          KU178539.1 - 4, 5 991

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          protein heterooligomerization
          cell division
          cell-cell adhesion
          positive regulation of nonmotile primary cilium assembly
          GTPase activity
          protein binding
          GTP binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0