Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC24A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252231.1 | NP_001239160.1 | 3 | 900 | ||
NM_021982.2 | NP_068817.1 | 3 | 900 | |||
XM_017008961.1 | XP_016864450.1 | 3 | 545 | |||
XM_017008962.1 | XP_016864451.1 | 2 | 205 | |||
GenBank mRNA | AJ131244.1 | - | 3 | 563 | ||
AK304060.1 | - | 3 | 900 | |||
BC019341.1 | - | 3 | 861 | 3355949 |