Search
Search

HPSE| Gene Symbol: HPSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098540.2 | NP_001092010.1 | 12 | 1603 | ||
| NM_001166498.2 | NP_001159970.1 | 11 | 1419 | |||
| NM_001199830.1 | NP_001186759.1 | 11 | 1361 | |||
| NM_006665.5 | NP_006656.2 | 13 | 1641 | |||
| GenBank mRNA | AF084467.1 | - | 12 | 1510 | ||
| AF144325.1 | - | 12 | 1603 | |||
| AF152376.1 | - | 12 | 1518 | |||
| AF155510.1 | - | 13 | 1640 | |||
| AF165154.1 | - | 12 | 1555 | |||
| AK075400.1 | - | 12 | 1567 | |||
| AK222986.1 | - | 12 | 1569 | |||
| AM419200.1 | - | 11 | 1330 | |||
| AY948074.1 | - | 12 | 1509 | |||
| BC051321.1 | - | 13 | 1641 | 6250265 | ||
| FJ517659.1 | - | 12 | 1472 | |||
| FJ517660.1 | - | 11 | 1298 | |||
| GQ337901.1 | - | 11 | 1419 | |||
| GQ337902.1 | - | 12 | 1526 | |||