Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201550.2 | NP_001188479.1 | 2 | 368 | ||
NM_001201551.1 | NP_001188480.1 | 2 | 365 | |||
NM_006684.4 | NP_006675.2 | 2 | 368 | |||
XM_006711129.3 | XP_006711192.1 | 2 | 307 | |||
XM_017000110.1 | XP_016855599.1 | 2 | 331 | |||
XM_017000111.1 | XP_016855600.1 | 2 | 331 | |||
XM_017000112.1 | XP_016855601.1 | 2 | 331 | |||
XM_017000113.1 | XP_016855602.1 | 2 | 331 | |||
GenBank mRNA | AJ640130.2 | - | 2 | 297 | ||
AK292752.1 | - | 2 | 365 |
Gene Symbol: CFHR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG618529.1 | - | 2 | 319 |