Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPARGC1A SIRNA Assays ›
          Gene Symbol:
          PPARGC1A
          Gene Name:
          PPARG coactivator 1 alpha
          Gene Aliases:
          LEM6, PGC-1(alpha), PGC-1alpha, PGC-1v, PGC1, PGC1A, PPARGC1
          Chromosome Location:
          Chr.4: 23792021 - 24472975 on Build GRCh38
          UniGene ID:
          Hs.527078
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s21394

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPARGC1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013261.3 NP_037393.1 8 1655
          XM_005248131.4 XP_005248188.1 8 2398
          XM_005248132.1 XP_005248189.1 8 1577
          XM_005248134.4 XP_005248191.1 8 1644
          XM_011513765.2 XP_011512067.1 9 2147
          XM_011513766.1 XP_011512068.1 9 1729
          XM_011513767.2 XP_011512069.1 9 3867
          XM_011513768.1 XP_011512070.1 8 1628
          XM_011513769.2 XP_011512071.1 8 1644
          XM_011513770.2 XP_011512072.1 7 1865
          XM_011513771.1 XP_011512073.1 7 1474
          XM_017007664.1 XP_016863153.1 10 2104
          GenBank mRNA AB061325.1 - 8 1641
          AF106698.1 - 8 1655
          AF159714.1 - 8 1564
          AF186379.1 - 8 1574
          HQ695733.1 - 7 1865
          JQ772116.1 - 9 1943
          JQ772117.1 - 9 2046
          JQ772118.1 - 10 2142
          JQ772119.1 - 10 2147
          JQ772120.1 - 10 2229

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          mitophagy
          temperature homeostasis
          cellular glucose homeostasis
          negative regulation of protein phosphorylation
          response to dietary excess
          response to ischemia
          galactose metabolic process
          gluconeogenesis
          regulation of transcription, DNA-templated
          transcription initiation from RNA polymerase II promoter
          mRNA processing
          protein complex assembly
          mitochondrion organization
          positive regulation of transcription from RNA polymerase II promoter by pheromones
          aging
          digestion
          circadian rhythm
          androgen metabolic process
          RNA splicing
          response to cold
          positive regulation of mitochondrion organization
          skeletal muscle atrophy
          response to muscle activity
          response to electrical stimulus involved in regulation of muscle adaptation
          negative regulation of smooth muscle cell migration
          fatty acid oxidation
          cerebellum development
          respiratory electron transport chain
          androgen receptor signaling pathway
          forebrain development
          circadian regulation of gene expression
          cellular response to oxidative stress
          positive regulation of histone acetylation
          cellular response to potassium ion
          response to statin
          response to drug
          response to starvation
          regulation of circadian rhythm
          response to leucine
          negative regulation of neuron apoptotic process
          cellular respiration
          positive regulation of gluconeogenesis
          negative regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          positive regulation of smooth muscle cell proliferation
          negative regulation of smooth muscle cell proliferation
          protein stabilization
          brown fat cell differentiation
          positive regulation of sequence-specific DNA binding transcription factor activity
          flavone metabolic process
          adipose tissue development
          cellular response to lipopolysaccharide
          cellular response to nitrite
          cellular response to caffeine
          cellular response to fructose stimulus
          cellular response to glucose stimulus
          cellular response to interleukin-6
          cellular response to tumor necrosis factor
          cellular response to follicle-stimulating hormone stimulus
          cellular response to estradiol stimulus
          cellular response to hypoxia
          cellular response to transforming growth factor beta stimulus
          response to epinephrine
          response to norepinephrine
          negative regulation of mitochondrial fission
          cellular response to thyroid hormone stimulus
          negative regulation of neuron death
          response to metformin
          positive regulation of cellular respiration
          positive regulation of mitochondrial DNA metabolic process
          positive regulation of muscle tissue development
          positive regulation of glomerular visceral epithelial cell apoptotic process
          cellular response to ionomycin
          cellular response to resveratrol
          response to methionine
          adaptive thermogenesis
          positive regulation of progesterone biosynthetic process
          negative regulation of receptor activity
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of energy homeostasis
          positive regulation of ATP biosynthetic process
          nucleotide binding
          RNA polymerase II transcription cofactor activity
          DNA binding
          transcription coactivator activity
          RNA binding
          protein binding
          transcription factor binding
          ligand-dependent nuclear receptor binding
          estrogen receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          chromatin DNA binding
          ubiquitin protein ligase binding
          peroxisome proliferator activated receptor binding
          alpha-tubulin binding
          sequence-specific DNA binding
          androgen receptor binding
          promoter-specific chromatin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0