Search
Search

MMP24| Gene Symbol: MMP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006690.3 | NP_006681.1 | 7, 8 | 1337 | ||
| XM_017027597.1 | XP_016883086.1 | 6, 7 | 1172 | |||
| XM_017027598.1 | XP_016883087.1 | 7, 8 | 1308 | |||
| GenBank mRNA | AB021227.1 | - | 7, 8 | 1337 | ||
| AF131284.1 | - | 7, 8 | 1334 | |||
| BC047614.1 | - | 7, 8 | 1277 | |||
| DQ300264.1 | - | 8, 9 | 1178 | |||