Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006710.4 | NP_006701.1 | 3 | 708 | ||
NM_198189.2 | NP_937832.1 | 4 | 773 | |||
GenBank mRNA | AI690743.1 | - | 3 | 437 | ||
AK289891.1 | - | 4 | 307 | |||
AK294078.1 | - | 3 | 263 | |||
AK308962.1 | - | 3 | 235 | |||
AK313437.1 | - | 3 | 251 | |||
BC003090.1 | - | 3 | 232 | 3503287 | ||
BC036499.1 | - | 3 | 842 | 5264527 | ||
BC080617.1 | - | 3 | 204 | 6252628 | ||
BF979628.1 | - | 337 | ||||
BG721393.1 | - | 4 | 311 | |||
CR456994.1 | - | 3 | 184 | |||
U51205.1 | - | 3 | 233 |