Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRDX3 SIRNA Assays ›
          Gene Symbol:
          PRDX3
          Gene Name:
          peroxiredoxin 3
          Gene Aliases:
          AOP-1, AOP1, HBC189, MER5, PRO1748, SP-22, prx-III
          Chromosome Location:
          Chr.10: 119167699 - 119178865 on Build GRCh38
          UniGene ID:
          Hs.523302
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s21509
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRDX3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001302272.1 NP_001289201.1 4 430
          NM_006793.4 NP_006784.1 4 430
          NR_126102.1 3 297
          NR_126103.1 2 155
          NR_126105.1 3 147
          GenBank mRNA AK130897.1 - 4 377
          AK222659.1 - 4 360
          AK313169.1 - 4 368
          AV718278.1 - 4 327
          AV761690.1 - 3 243
          BC002685.1 - 4 361 3607558
          BC007062.1 - 4 385 3960644
          BC008435.1 - 4 369 4081846
          BC009601.1 - 4 362 4134666
          BC021691.1 - 4 402 3961379
          BC022373.1 - 4 375 4775243
          BC059169.1 - 4 362 4133576
          BC111397.1 - 4 369 4246143
          BF209381.1 - 3 86
          BG115645.1 - 2 79
          BG282768.1 - 4 344
          BT020007.1 - 4 355
          BX382389.2 - 4 379
          CD687044.1 - 4 401
          CR450344.1 - 4 355
          D49396.1 - 4 361
          EF036492.1 - 3 330
          HY173455.1 - 4 430

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00428953_g1

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          maternal placenta development
          apoptotic process
          response to oxidative stress
          mitochondrion organization
          positive regulation of cell proliferation
          peptidyl-cysteine oxidation
          myeloid cell differentiation
          response to lipopolysaccharide
          negative regulation of kinase activity
          cellular response to oxidative stress
          cellular response to reactive oxygen species
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          cell redox homeostasis
          positive regulation of NF-kappaB transcription factor activity
          regulation of mitochondrial membrane potential
          cellular oxidant detoxification
          protein binding
          protein C-terminus binding
          thioredoxin peroxidase activity
          alkyl hydroperoxide reductase activity
          kinase binding
          protein kinase binding
          identical protein binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0