Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197233.2 | NP_001184162.1 | 4 | 947 | ||
NM_001197234.2 | NP_001184163.1 | 5 | 1059 | |||
NM_007049.4 | NP_008980.1 | 5 | 1059 | |||
NM_078476.3 | NP_510961.1 | 5 | 1059 | |||
GenBank mRNA | AB209777.1 | - | 5 | 995 | ||
AK297098.1 | - | 4 | 901 | |||
BC016661.1 | - | 5 | 1002 | 4340392 | ||
BX538050.1 | - | 4 | 602 | |||
U90543.1 | - | 5 | 1000 |