siRNA Details
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030990.2 | NP_112252.1 | 5, 6 | 843 | ||
XM_006257293.2 | XP_006257355.1 | 8, 9 | 1125 | |||
XM_008773401.1 | XP_008771623.1 | 8, 9 | 1229 | |||
GenBank mRNA | BC093596.1 | - | 4, 5 | 704 | 7314758 | |
FQ211530.1 | - | 5, 6 | 816 | |||
FQ212519.1 | - | 4, 5 | 452 | |||
FQ212680.1 | - | 4, 5 | 557 | |||
FQ213540.1 | - | 4, 5 | 557 | |||
FQ213548.1 | - | 4, 5 | 557 | |||
FQ213841.1 | - | 4, 5 | 557 | |||
FQ213943.1 | - | 4, 5 | 557 | |||
FQ214012.1 | - | 4, 5 | 557 | |||
FQ214050.1 | - | 4, 5 | 557 | |||
FQ214153.1 | - | 4, 5 | 558 | |||
FQ214556.1 | - | 4, 5 | 557 | |||
M11185.1 | - | 5, 6 | 818 | |||
M14126.1 | - | 2, 3 | 171 | |||
M25888.1 | - | 3, 4 | 383 | |||
X02809.1 | - | 5, 6 | 747 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
