Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013448.2 | NP_038476.2 | 22 | 3973 | ||
NM_182648.1 | NP_872589.1 | 21 | 3877 | |||
XM_011536374.1 | XP_011534676.1 | 21 | 3272 | |||
XM_011536376.2 | XP_011534678.2 | 12 | 2100 | |||
XM_017020942.1 | XP_016876431.1 | 11 | 2003 | |||
GenBank mRNA | AB032252.1 | - | 21 | 3874 | ||
AB209255.1 | - | 13 | 2224 | |||
AF213467.1 | - | 22 | 3571 | |||
AF221130.1 | - | 21 | 3405 | |||
AK027289.1 | - | 2 | 219 | |||
AL050089.1 | - | 5 | 1170 |