Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fopnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108261.1 | NP_001101731.1 | 3 | 349 | ||
XM_006245848.2 | XP_006245910.1 | 3 | 378 | |||
GenBank mRNA | BC083733.1 | - | 3 | 323 | ||
BC158662.1 | - | 3 | 315 | 8369763 | ||
FQ215640.1 | - | 3 | 315 | |||
FQ228774.1 | - | 3 | 323 | |||
FQ228837.1 | - | 3 | 305 |