Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HBB-B1 SIRNA Assays ›
          Gene Symbol:
          Hbb-b1 Hbb LOC100134871 LOC103694855
          Gene Name:
          hemoglobin, beta adult major chain
          hemoglobin, beta
          beta globin minor gene
          hemoglobin subunit beta-2-like
          Gene Aliases:
          -
          -
          -
          -
          Chromosome Location:
          Chr.1: 168945529 - 168953023 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 168971269 - 168972680 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 168957460 - 168958836 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906791: 23525 - 31030 on Build Rnor_6.0
          UniGene ID:
          Rn.36966
          Rn.202945
          Rn.216394
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s221390

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hbb-b1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_198776.1 NP_942071.1 3 583
          GenBank mRNA BC058502.1 - 3 583 6887460
          FQ218177.1 - 3 577
          Gene Symbol: Hbb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033234.1 NP_150237.1 3 574
          GenBank mRNA BC058448.1 - 3 583 6921704
          FQ209466.1 - 3 579
          FQ209544.1 - 3 579
          FQ209725.1 - 3 581
          FQ209747.1 - 3 579
          FQ209754.1 - 3 579
          FQ209760.1 - 3 579
          FQ209781.1 - 3 579
          FQ209789.1 - 3 579
          FQ209796.1 - 3 579
          FQ209806.1 - 3 581
          FQ209822.1 - 579
          FQ209826.1 - 3 588
          FQ209841.1 - 3 581
          FQ209870.1 - 3 581
          FQ209871.1 - 3 579
          FQ209903.1 - 3 579
          FQ209929.1 - 3 579
          FQ209937.1 - 3 579
          FQ209954.1 - 3 579
          FQ209961.1 - 3 578
          FQ209996.1 - 3 579
          FQ210038.1 - 3 581
          FQ210054.1 - 3 579
          FQ210058.1 - 3 579
          FQ210090.1 - 3 581
          FQ210123.1 - 3 579
          FQ210127.1 - 3 579
          FQ210137.1 - 3 579
          FQ210175.1 - 3 577
          FQ210181.1 - 3 581
          FQ210200.1 - 3 579
          FQ210211.1 - 3 579
          FQ210216.1 - 3 578
          FQ210218.1 - 3 579
          FQ210280.1 - 3 602
          FQ210343.1 - 3 579
          FQ210348.1 - 3 579
          FQ210353.1 - 3 579
          FQ210360.1 - 3 579
          FQ210373.1 - 3 579
          FQ210415.1 - 3 579
          FQ210449.1 - 3 579
          FQ210466.1 - 3 579
          FQ210482.1 - 3 579
          FQ210486.1 - 3 581
          FQ210497.1 - 3 580
          FQ210499.1 - 3 579
          FQ210511.1 - 3 579
          FQ210523.1 - 3 579
          FQ210534.1 - 3 579
          FQ210558.1 - 3 579
          FQ210602.1 - 3 579
          FQ210676.1 - 3 579
          FQ210677.1 - 3 579
          FQ210735.1 - 3 578
          FQ210771.1 - 3 579
          FQ210783.1 - 3 571
          FQ210789.1 - 3 579
          FQ210794.1 - 3 579
          FQ210806.1 - 3 579
          FQ210826.1 - 3 579
          FQ210828.1 - 3 576
          FQ210829.1 - 3 579
          FQ210838.1 - 3 579
          FQ210852.1 - 3 579
          FQ210875.1 - 3 579
          FQ210912.1 - 3 579
          FQ210937.1 - 3 579
          FQ210949.1 - 3 582
          FQ210960.1 - 3 579
          FQ210984.1 - 3 579
          FQ210985.1 - 3 579
          FQ210998.1 - 3 579
          FQ211014.1 - 3 579
          FQ211016.1 - 3 579
          FQ211034.1 - 3 579
          FQ211061.1 - 3 578
          FQ211065.1 - 3 579
          FQ211103.1 - 3 579
          FQ211125.1 - 3 580
          FQ211133.1 - 3 576
          FQ211137.1 - 3 579
          FQ211140.1 - 3 579
          FQ211147.1 - 3 579
          FQ211154.1 - 3 579
          FQ211182.1 - 3 579
          FQ211193.1 - 3 579
          FQ211207.1 - 3 581
          FQ211241.1 - 3 579
          FQ211242.1 - 3 579
          FQ211290.1 - 3 578
          FQ211314.1 - 3 579
          FQ211331.1 - 3 579
          FQ211336.1 - 3 579
          FQ211340.1 - 3 579
          FQ211423.1 - 3 581
          FQ211443.1 - 3 579
          FQ211452.1 - 3 579
          FQ211477.1 - 3 579
          FQ211497.1 - 3 579
          FQ211548.1 - 3 579
          FQ211583.1 - 3 579
          FQ211590.1 - 3 580
          FQ211613.1 - 3 581
          FQ211617.1 - 3 579
          FQ211626.1 - 3 579
          FQ211692.1 - 3 579
          FQ211696.1 - 3 579
          FQ211790.1 - 3 578
          FQ211806.1 - 3 579
          FQ211815.1 - 3 579
          FQ211829.1 - 3 581
          FQ211847.1 - 3 579
          FQ211858.1 - 3 579
          FQ211864.1 - 3 579
          FQ211905.1 - 3 579
          FQ211931.1 - 3 579
          FQ212127.1 - 3 579
          FQ212156.1 - 3 577
          FQ212234.1 - 3 579
          FQ212241.1 - 3 579
          FQ212258.1 - 3 578
          FQ212275.1 - 3 579
          FQ212287.1 - 3 579
          FQ212303.1 - 3 579
          FQ212341.1 - 3 581
          FQ212449.1 - 3 579
          FQ213270.1 - 3 579
          FQ213352.1 - 3 579
          FQ213466.1 - 3 579
          FQ213534.1 - 3 579
          FQ213582.1 - 1058
          FQ213796.1 - 3 579
          FQ214044.1 - 3 579
          FQ214573.1 - 3 579
          FQ216995.1 - 3 581
          FQ217014.1 - 3 581
          FQ217108.1 - 3 579
          FQ217151.1 - 3 579
          FQ217201.1 - 3 579
          FQ217206.1 - 3 579
          FQ217586.1 - 3 581
          FQ217619.1 - 579
          FQ217623.1 - 3 579
          FQ217653.1 - 3 579
          FQ218033.1 - 3 578
          FQ218042.1 - 3 579
          FQ218100.1 - 3 579
          FQ218112.1 - 3 579
          FQ218118.1 - 3 581
          FQ218119.1 - 3 578
          FQ218124.1 - 3 583
          FQ218145.1 - 3 579
          FQ218171.1 - 3 579
          FQ218180.1 - 3 579
          FQ218182.1 - 3 579
          FQ218199.1 - 3 582
          FQ218229.1 - 3 582
          FQ218253.1 - 3 579
          FQ218272.1 - 3 576
          FQ218288.1 - 3 579
          FQ218461.1 - 3 599
          FQ218539.1 - 3 578
          FQ218748.1 - 3 579
          FQ218761.1 - 3 579
          FQ218763.1 - 3 579
          FQ218779.1 - 3 577
          FQ219114.1 - 3 578
          FQ219985.1 - 3 528
          FQ220085.1 - 3 579
          FQ220253.1 - 3 528
          FQ221076.1 - 3 579
          FQ221202.1 - 3 581
          FQ221246.1 - 3 579
          FQ221520.1 - 3 579
          FQ221738.1 - 3 578
          FQ221956.1 - 3 579
          FQ221962.1 - 3 579
          FQ222032.1 - 3 579
          FQ222262.1 - 3 579
          FQ222275.1 - 3 579
          FQ222291.1 - 3 579
          FQ222503.1 - 3 579
          FQ222804.1 - 3 579
          FQ222960.1 - 3 579
          FQ223002.1 - 3 579
          FQ223079.1 - 3 579
          FQ223164.1 - 3 576
          FQ223490.1 - 3 579
          FQ223631.1 - 3 579
          FQ223717.1 - 3 579
          FQ223745.1 - 3 581
          FQ223805.1 - 3 579
          FQ223852.1 - 3 571
          FQ223888.1 - 3 579
          FQ223889.1 - 3 581
          FQ223995.1 - 3 552
          FQ224088.1 - 3 579
          FQ224309.1 - 3 579
          FQ224343.1 - 3 579
          FQ224371.1 - 3 579
          FQ224406.1 - 3 579
          FQ224475.1 - 3 579
          FQ224682.1 - 3 579
          FQ224727.1 - 3 579
          FQ224732.1 - 3 579
          FQ225124.1 - 3 641
          FQ225436.1 - 3 579
          FQ225457.1 - 3 579
          FQ225469.1 - 3 579
          FQ225684.1 - 731
          FQ225738.1 - 3 579
          FQ225754.1 - 3 578
          FQ225816.1 - 3 579
          FQ226188.1 - 3 577
          FQ226536.1 - 3 579
          FQ226572.1 - 3 579
          FQ226889.1 - 3 579
          FQ227120.1 - 3 579
          FQ227498.1 - 3 579
          FQ227822.1 - 3 579
          FQ227875.1 - 3 579
          FQ228016.1 - 3 579
          FQ228244.1 - 3 579
          FQ228281.1 - 3 579
          FQ228294.1 - 3 579
          FQ228295.1 - 3 579
          FQ228296.1 - 3 581
          FQ228414.1 - 3 579
          FQ228616.1 - 3 579
          FQ228638.1 - 3 579
          FQ229015.1 - 3 579
          FQ229033.1 - 3 579
          FQ229489.1 - 3 579
          FQ230471.1 - 3 579
          FQ230486.1 - 3 577
          FQ230503.1 - 3 579
          FQ230508.1 - 3 579
          FQ230541.1 - 3 579
          FQ230861.1 - 3 579
          FQ230866.1 - 3 579
          FQ230889.1 - 3 579
          FQ231191.1 - 3 578
          FQ231208.1 - 3 578
          FQ231253.1 - 3 579
          FQ231286.1 - 3 579
          FQ231289.1 - 3 579
          FQ232655.1 - 3 611
          FQ233311.1 - 3 654
          FQ234713.1 - 3 581
          FQ234942.1 - 3 577
          FQ235254.1 - 1215
          M17084.1 - 3 531
          X16417.1 - 3 574
          Gene Symbol: LOC100134871
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113223.1 NP_001106694.1 3 578
          GenBank mRNA FQ209951.1 - 3 581
          FQ210023.1 - 3 581
          FQ210060.1 - 3 579
          FQ210106.1 - 3 579
          FQ210483.1 - 3 579
          FQ210600.1 - 3 581
          FQ210669.1 - 3 575
          FQ211039.1 - 3 573
          FQ211266.1 - 3 579
          FQ211272.1 - 3 579
          FQ211275.1 - 3 579
          FQ211294.1 - 3 581
          FQ211362.1 - 3 579
          FQ211415.1 - 3 579
          FQ211424.1 - 3 579
          FQ211697.1 - 3 581
          FQ212253.1 - 3 579
          FQ212317.1 - 3 581
          FQ212406.1 - 3 581
          FQ212421.1 - 3 579
          FQ213080.1 - 3 608
          FQ214057.1 - 3 579
          FQ214129.1 - 3 579
          FQ217131.1 - 3 577
          FQ217673.1 - 3 579
          FQ218002.1 - 3 580
          FQ218191.1 - 3 579
          FQ218246.1 - 3 551
          FQ218266.1 - 3 617
          FQ218689.1 - 3 579
          FQ218821.1 - 3 579
          FQ219109.1 - 3 579
          FQ219133.1 - 3 610
          FQ221588.1 - 3 577
          FQ222844.1 - 3 579
          FQ224149.1 - 3 579
          FQ224301.1 - 3 579
          FQ224598.1 - 3 579
          FQ224632.1 - 3 579
          FQ224738.1 - 3 579
          FQ224763.1 - 3 618
          FQ225377.1 - 3 579
          FQ225739.1 - 3 581
          FQ226460.1 - 3 579
          FQ226535.1 - 3 579
          FQ226609.1 - 3 581
          FQ228555.1 - 3 579
          FQ230881.1 - 3 581
          FQ232298.1 - 723
          FQ232442.1 - 3 610
          FQ233001.1 - 3 579
          X05080.1 - 3 578
          X16418.1 - 3 544
          Gene Symbol: LOC103694855
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773799.1 XP_008772021.1 3 619

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          oxygen transport
          glutathione metabolic process
          regulation of eIF2 alpha phosphorylation by heme
          hemopoiesis
          erythrocyte development
          protein heterooligomerization
          positive regulation of cell death
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          oxidation-reduction process
          renal absorption
          platelet aggregation
          oxygen transporter activity
          iron ion binding
          oxygen binding
          heme binding
          hemoglobin alpha binding
          hemoglobin beta binding
          peroxidase activity
          hemoglobin binding
          haptoglobin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0