Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53inp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270947.1 | NP_001257876.1 | 3 | 563 | ||
XM_003749598.3 | XP_003749646.1 | 3 | 701 | |||
XM_006235327.2 | XP_006235389.1 | 3 | 1275 | |||
XM_006235329.2 | XP_006235391.1 | 3 | 1469 | |||
GenBank mRNA | AJ297794.1 | - | 3 | 513 |