Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loh12cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108650.2 | NP_001102120.1 | 2 | 117 | ||
XM_006237528.2 | XP_006237590.1 | 2 | 850 | |||
XM_008763394.1 | XP_008761616.1 | 2 | 848 | |||
XM_008763395.1 | XP_008761617.1 | 2 | 848 | |||
XM_008763396.1 | XP_008761618.1 | 2 | 850 | |||
GenBank mRNA | BC160872.1 | - | 2 | 117 | 7365768 | |
FQ211769.1 | - | 2 | 212 |