Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108813.2 | NP_001102283.2 | 1, 2 | 196 | ||
GenBank mRNA | BC168213.1 | - | 1, 2 | 172 | ||
CB567280.1 | - | 1, 2 | 213 | |||
FM054992.1 | - | 1, 2 | 181 | |||
FQ214979.1 | - | 1, 2 | 157 |
Gene Symbol: NEWGENE_1560955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773489.1 | XP_008771711.1 | 1, 2 | 287 |