Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008971.1 | NP_001008971.1 | 1 | 118 | ||
XM_006236848.1 | XP_006236910.1 | 3 | 454 | |||
GenBank mRNA | AY510293.1 | - | 1 | 118 | ||
U36896.1 | - | 3 | 468 |
Gene Symbol: Vom1r92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008968.1 | NP_001008968.1 | 1 | 112 | ||
XM_006236847.1 | XP_006236909.1 | 2 | 187 | |||
GenBank mRNA | AY510279.1 | - | 1 | 112 |