Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031706.1 | NP_113894.1 | 6 | 622 | ||
GenBank mRNA | BC082802.1 | - | 5 | 606 | 7302521 | |
BC166878.1 | - | 6 | 622 | 6920981 | ||
FQ221315.1 | - | 6 | 622 | |||
FQ222655.1 | - | 6 | 659 | |||
FQ222951.1 | - | 6 | 620 | |||
FQ223011.1 | - | 6 | 622 | |||
FQ223286.1 | - | 6 | 624 | |||
FQ229267.1 | - | 6 | 621 | |||
X06423.1 | - | 6 | 622 |