Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNRG SIRNA Assays ›
Gene Symbol:
SYNRG
Gene Name:
synergin, gamma
Gene Aliases:
AP1GBP1, SYNG
Chromosome Location:
Chr.17: 37514797 - 37613637 on Build GRCh38
UniGene ID:
Hs.594647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22267
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNRG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163544.2
NP_001157016.1
8
874
NM_001163545.2
NP_001157017.1
8
871
NM_001163546.2
NP_001157018.1
8
874
NM_001163547.2
NP_001157019.1
8
874
NM_007247.5
NP_009178.3
9
1108
NM_080550.4
NP_542117.3
8
874
NM_198882.2
NP_942583.1
8
874
XM_005256980.4
XP_005257037.1
10
1469
XM_017024076.1
XP_016879565.1
10
1469
XM_017024077.1
XP_016879566.1
10
1388
XM_017024078.1
XP_016879567.1
10
1469
XM_017024079.1
XP_016879568.1
10
1469
XM_017024080.1
XP_016879569.1
10
5521
XM_017024081.1
XP_016879570.1
10
5518
XM_017024082.1
XP_016879571.1
10
1469
XM_017024083.1
XP_016879572.1
10
1469
XM_017024084.1
XP_016879573.1
10
1469
XM_017024085.1
XP_016879574.1
10
1469
XM_017024086.1
XP_016879575.1
10
1469
XM_017024087.1
XP_016879576.1
10
1469
XM_017024088.1
XP_016879577.1
9
1235
XM_017024089.1
XP_016879578.1
10
1469
XM_017024090.1
XP_016879579.1
9
1166
XM_017024091.1
XP_016879580.1
9
1085
XM_017024092.1
XP_016879581.1
10
1469
XM_017024093.1
XP_016879582.1
9
1166
XM_017024094.1
XP_016879583.1
9
1085
XM_017024095.1
XP_016879584.1
9
1166
XM_017024096.1
XP_016879585.1
10
1469
XM_017024097.1
XP_016879586.1
9
1166
XM_017024098.1
XP_016879587.1
9
1166
XM_017024099.1
XP_016879588.1
8
851
XM_017024100.1
XP_016879589.1
9
1166
XM_017024101.1
XP_016879590.1
9
1085
XM_017024102.1
XP_016879591.1
8
851
XM_017024103.1
XP_016879592.1
8
932
XM_017024104.1
XP_016879593.1
8
932
XM_017024105.1
XP_016879594.1
10
1469
XM_017024106.1
XP_016879595.1
8
932
GenBank mRNA
AF169548.1
-
9
1068
AK025335.1
-
2
264
AK126988.1
-
9
1836
AK302821.1
-
10
1346
AK308292.1
-
3
487
BC090930.1
-
8
825
3077104
BC117313.1
-
8
814
40125864
BC143476.1
-
8
817
9051984
BC143478.1
-
8
817
9051986
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053419, EF121981, EF121979, EF121980, EF121978, and others.
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
endocytosis
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0