Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010938.1 | NP_001010938.1 | 2 | 443 | ||
NM_001308046.1 | NP_001294975.1 | 2 | 501 | |||
NM_005781.4 | NP_005772.3 | 2 | 652 | |||
XM_005269268.3 | XP_005269325.1 | 2 | 443 | |||
XM_005269270.3 | XP_005269327.1 | 2 | 291 | |||
XM_005269275.3 | XP_005269332.1 | 2 | 465 | |||
XM_011512317.2 | XP_011510619.1 | 4 | 1010 | |||
XM_011512318.2 | XP_011510620.2 | 2 | 382 | |||
XM_011512320.1 | XP_011510622.1 | 2 | 165 | |||
XM_017005508.1 | XP_016860997.1 | 2 | 382 | |||
XM_017005509.1 | XP_016860998.1 | 2 | 382 | |||
XM_017005510.1 | XP_016860999.1 | 2 | 382 | |||
GenBank mRNA | AK131539.1 | - | 2 | 443 | ||
AK225786.1 | - | 2 | 448 | |||
AK309332.1 | - | 2 | 279 | |||
BC008884.2 | - | 2 | 298 | |||
BC020389.1 | - | 2 | 492 | |||
BC028164.1 | - | 2 | 166 | |||
L13738.2 | - | 2 | 727 |