Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008392.1 | NP_001008393.1 | 4 | 305 | ||
NM_005808.2 | NP_005799.2 | 3 | 272 | |||
XM_017005519.1 | XP_016861008.1 | 4 | 331 | |||
XM_017005520.1 | XP_016861009.1 | 4 | 1649 | |||
GenBank mRNA | AI143812.1 | - | 4 | 305 | ||
AI192993.1 | - | 3 | 272 | |||
AJ575644.1 | - | 3 | 272 | |||
AJ575645.1 | - | 4 | 305 | |||
AY279532.1 | - | 3 | 250 | |||
AY364238.1 | - | 3 | 250 | |||
D88153.1 | - | 4 | 622 |