Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAMKK2 SIRNA Assays ›
          Gene Symbol:
          CAMKK2
          Gene Name:
          calcium/calmodulin dependent protein kinase kinase 2
          Gene Aliases:
          CAMKK, CAMKKB
          Chromosome Location:
          Chr.12: 121237692 - 121298308 on Build GRCh38
          UniGene ID:
          Hs.297343
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s223143

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAMKK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270485.1 NP_001257414.1 4, 5 741
          NM_001270486.1 NP_001257415.1 3, 4 625
          NM_006549.3 NP_006540.3 4, 5 1395
          NM_153499.2 NP_705719.2 4, 5 1395
          NM_153500.1 NP_705720.1 4, 5 1395
          NM_172214.2 NP_757363.1 4, 5 1395
          NM_172215.2 NP_757364.1 4, 5 1395
          NM_172216.1 NP_757365.1 4, 5 1395
          NM_172226.2 NP_757380.1 4, 5 741
          XM_005253822.3 XP_005253879.1 4, 5 697
          XM_005253823.1 XP_005253880.1 4, 5 697
          XM_005253824.3 XP_005253881.1 4, 5 697
          XM_011537763.1 XP_011536065.1 4, 5 697
          XM_011537764.2 XP_011536066.1 4, 5 743
          XM_017018694.1 XP_016874183.1 5, 6 751
          XM_017018695.1 XP_016874184.1 5, 6 960
          XM_017018696.1 XP_016874185.1 5, 6 802
          XM_017018697.1 XP_016874186.1 5, 6 824
          XM_017018698.1 XP_016874187.1 4, 5 748
          XM_017018699.1 XP_016874188.1 5, 6 751
          XM_017018700.1 XP_016874189.1 5, 6 751
          XM_017018701.1 XP_016874190.1 4, 5 748
          XM_017018702.1 XP_016874191.1 4, 5 906
          XM_017018703.1 XP_016874192.1 5, 6 751
          XM_017018704.1 XP_016874193.1 4, 5 748
          XM_017018705.1 XP_016874194.1 5, 6 751
          XM_017018706.1 XP_016874195.1 4, 5 743
          XM_017018707.1 XP_016874196.1 3, 4 666
          XM_017018708.1 XP_016874197.1 4, 5 743
          XM_017018709.1 XP_016874198.1 4, 5 700
          GenBank mRNA AB018330.2 - 3, 4 571
          AB081336.1 - 3, 4 598
          AB081337.1 - 3, 4 598
          AF101264.1 - 3, 4 218
          AF140507.1 - 4, 5 679
          AF287630.1 - 4, 5 1389
          AF287631.1 - 4, 5 1389
          AF321385.1 - 4, 5 1145
          AF321386.1 - 4, 5 1145
          AF321387.1 - 4, 5 1145
          AF321388.1 - 4, 5 1145
          AK024748.1 - 3, 4 381
          AK292072.1 - 4, 5 889
          AK300865.1 - 5, 6 641
          BC026060.2 - 4, 5 674 3842706

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein phosphorylation
          peptidyl-tyrosine phosphorylation
          calcium-mediated signaling
          regulation of protein kinase activity
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          calmodulin-dependent protein kinase activity
          protein tyrosine kinase activity
          calcium ion binding
          calmodulin binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0