Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001865.3 | NP_001856.2 | 4 | 500 | ||
NR_029466.1 | 4 | 553 | ||||
GenBank mRNA | CB139775.1 | - | 4 | 412 | ||
CK821341.1 | - | 4 | 511 | |||
HE647861.1 | - | 4 | 500 | |||
X15822.1 | - | 4 | 319 |