Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DRD3 SIRNA Assays ›
          Gene Symbol:
          DRD3
          Gene Name:
          dopamine receptor D3
          Gene Aliases:
          D3DR, ETM1, FET1
          Chromosome Location:
          Chr.3: 114127623 - 114199407 on Build GRCh38
          UniGene ID:
          Hs.121478
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s223439

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DRD3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000796.5 NP_000787.2 6 1179
          NM_001282563.2 NP_001269492.1 7 1038
          NM_001290809.1 NP_001277738.1 7 1171
          NM_033663.5 NP_387512.3 6 1179
          XM_017005829.1 XP_016861318.1 7 999
          GenBank mRNA AK292309.1 - 7 1038
          BC095510.1 - 5 749 7001986
          BC128122.1 - 7 1093 40112299
          BC128123.1 - 6 825 40112300
          KJ534829.1 - 5 748
          KJ535032.1 - 4 635
          L20469.1 - 5 748
          U32499.1 - 5 748

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          synaptic transmission, dopaminergic
          response to amphetamine
          regulation of blood volume by renin-angiotensin
          G-protein coupled receptor internalization
          cellular calcium ion homeostasis
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          negative regulation of adenylate cyclase activity
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          learning or memory
          learning
          locomotory behavior
          positive regulation of cell proliferation
          visual learning
          regulation of dopamine secretion
          regulation of lipid metabolic process
          regulation of cAMP metabolic process
          negative regulation of sodium:proton antiporter activity
          positive regulation of cytokinesis
          circadian regulation of gene expression
          response to histamine
          social behavior
          gastric emptying
          positive regulation of renal sodium excretion
          regulation of multicellular organism growth
          response to cocaine
          dopamine metabolic process
          response to drug
          response to morphine
          regulation of circadian sleep/wake cycle, sleep
          response to ethanol
          negative regulation of blood pressure
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          acid secretion
          behavioral response to cocaine
          negative regulation of oligodendrocyte differentiation
          arachidonic acid secretion
          negative regulation of protein secretion
          musculoskeletal movement, spinal reflex action
          regulation of dopamine uptake involved in synaptic transmission
          negative regulation of protein kinase B signaling
          prepulse inhibition
          negative regulation of dopamine receptor signaling pathway
          positive regulation of dopamine receptor signaling pathway
          regulation of locomotion involved in locomotory behavior
          dopamine neurotransmitter receptor activity, coupled via Gs
          dopamine neurotransmitter receptor activity, coupled via Gi/Go
          protein binding
          drug binding
          protein domain specific binding
          D1 dopamine receptor binding
          dopamine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5l6s2:80/100.66.78.182:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0