Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144831.1 | NP_001138303.1 | 8 | 1048 | ||
NM_001267700.1 | NP_001254629.1 | 7 | 934 | |||
GenBank mRNA | AF126021.1 | - | 8 | 1023 | ||
AF150962.1 | - | 8 | 961 | |||
AF178980.1 | - | 8 | 937 | |||
AF279895.1 | - | 3 | 149 | |||
AK298217.1 | - | 7 | 854 | |||
AK301316.1 | - | 7 | 1124 | |||
BC013228.1 | - | 8 | 931 | |||
BC014766.1 | - | 8 | 918 | 3452360 | ||
BC110322.1 | - | 8 | 1048 | 5540527 | ||
U72511.1 | - | 6 | 576 |