Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FCER1G SIRNA Assays ›
Gene Symbol:
FCER1G
Gene Name:
Fc fragment of IgE receptor Ig
Gene Aliases:
FCRG
Chromosome Location:
Chr.1: 161215297 - 161219248 on Build GRCh38
UniGene ID:
Hs.433300
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223520
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FCER1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004106.1
NP_004097.1
2, 3
170
GenBank mRNA
AK307102.1
-
2, 3
195
M33195.1
-
2, 3
170
Back To Top
More Information
Panther Classification:
Molecular Function -
defense/immunity protein
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of type IIa hypersensitivity
positive regulation of type III hypersensitivity
positive regulation of type I hypersensitivity
stimulatory C-type lectin receptor signaling pathway
neutrophil activation involved in immune response
T cell differentiation involved in immune response
Fc receptor mediated stimulatory signaling pathway
serotonin secretion by platelet
phagocytosis, engulfment
integrin-mediated signaling pathway
blood coagulation
regulation of platelet activation
immunoglobulin mediated immune response
antigen processing and presentation of exogenous peptide antigen via MHC class II
platelet activation
neutrophil chemotaxis
receptor internalization
positive regulation of interleukin-10 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of mast cell cytokine production
negative regulation of mast cell apoptotic process
Fc-gamma receptor signaling pathway
Fc-epsilon receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I
defense response to bacterium
positive regulation of mast cell degranulation
innate immune response
mast cell activation
positive regulation of phagocytosis
leukocyte migration
cellular response to low-density lipoprotein particle stimulus
protein localization to plasma membrane
protein binding
IgE receptor activity
IgE binding
IgG binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-q5zrm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline