Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FHL1 SIRNA Assays ›
Gene Symbol:
FHL1
LOC100128164
Gene Name:
four and a half LIM domains 1
four and a half LIM domains 1 pseudogene
Gene Aliases:
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
-
Chromosome Location:
Chr.X: 136146702 - 136211359 on Build GRCh38
Chromosome Location:
Chr.3: 169943984 - 169966734 on Build GRCh38
UniGene ID:
Hs.435369
Hs.633070
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s223534
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FHL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159699.1
NP_001153171.1
2, 3
425
NM_001159700.1
NP_001153172.1
3, 4
420
NM_001159701.1
NP_001153173.1
2, 3
296
NM_001159702.2
NP_001153174.1
3, 4
489
NM_001159703.1
NP_001153175.1
3, 4
489
NM_001159704.1
NP_001153176.1
2, 3
871
NM_001167819.1
NP_001161291.1
3, 4
357
NM_001449.4
NP_001440.2
3, 4
489
NR_027621.1
3, 4
571
XM_006724743.2
XP_006724806.1
2, 3
293
XM_006724744.2
XP_006724807.1
3, 4
371
XM_006724745.3
XP_006724808.1
3, 4
309
XM_006724746.2
XP_006724809.1
2, 3
244
XM_006724747.2
XP_006724810.1
2, 3
244
XM_011531316.1
XP_011529618.1
2, 3
293
XM_017029357.1
XP_016884846.1
3, 4
371
GenBank mRNA
AF063002.1
-
2, 3
244
AF098518.1
-
2, 3
213
AF220153.1
-
1, 2
160
AK122708.1
-
2, 3
872
AK223299.1
-
1, 2
194
AK289411.1
-
3, 4
571
AK299381.1
-
2, 3
271
AK299450.1
-
2, 3
268
AK300966.1
-
4, 5
296
AK301642.1
-
2, 3
293
AK304680.1
-
3, 4
348
BC010998.2
-
3, 4
368
4039973
BC088369.1
-
2, 3
258
30529295
BE965488.2
-
3, 4
297
CR456974.1
-
1, 2
160
DA497183.1
-
3, 4
489
DA917361.1
-
1, 2
55
DA982372.1
-
3, 4
358
DC363191.1
-
3, 4
420
DC373304.1
-
2, 3
297
DC373386.1
-
2, 3
426
U29538.1
-
1, 2
181
U60115.1
-
2, 3
243
Gene Symbol: LOC100128164
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027622.1
2
3426
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
muscle organ development
organ morphogenesis
negative regulation of G2/M transition of mitotic cell cycle
cell differentiation
negative regulation of cell growth
positive regulation of potassium ion transport
regulation of potassium ion transmembrane transporter activity
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
protein binding
zinc ion binding
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0