Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007348.3 | NP_031374.2 | 14 | 1741 | ||
XM_006711224.1 | XP_006711287.1 | 14 | 1662 | |||
XM_011509308.1 | XP_011507610.1 | 14 | 1722 | |||
XM_011509309.1 | XP_011507611.1 | 14 | 1719 | |||
XM_011509310.2 | XP_011507612.1 | 14 | 1722 | |||
GenBank mRNA | AB015856.1 | - | 14 | 1692 | ||
AB208929.1 | - | 14 | 1629 | |||
AF005887.1 | - | 14 | 1666 | |||
AK290498.1 | - | 14 | 1665 |