Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098482.1 | NP_001091952.1 | 6 | 636 | ||
NM_015321.2 | NP_056136.2 | 5 | 588 | |||
XM_005259833.3 | XP_005259890.1 | 6 | 597 | |||
XM_005259834.1 | XP_005259891.1 | 5 | 582 | |||
XM_005259835.3 | XP_005259892.1 | 6 | 596 | |||
XM_005259836.3 | XP_005259893.1 | 6 | 596 | |||
XM_006722710.3 | XP_006722773.1 | 6 | 596 | |||
XM_011527842.2 | XP_011526144.1 | 6 | 597 | |||
XM_017026536.1 | XP_016882025.1 | 6 | 597 | |||
XM_017026537.1 | XP_016882026.1 | 6 | 597 | |||
XM_017026538.1 | XP_016882027.1 | 6 | 597 | |||
GenBank mRNA | AB014516.1 | - | 6 | 501 | ||
AK024089.1 | - | 5 | 588 | |||
AU118537.1 | - | 5 | 588 | |||
AY040323.1 | - | 5 | 588 | |||
AY360171.1 | - | 6 | 573 | |||
BC017075.2 | - | 1 | 69 | |||
BC023614.2 | - | 5 | 411 | |||
BC028050.1 | - | 5 | 520 | 4938995 |