Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L13 SIRNA Assays ›
Gene Symbol:
BCL2L13
Gene Name:
BCL2 like 13
Gene Aliases:
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location:
Chr.22: 17628855 - 17730855 on Build GRCh38
UniGene ID:
Hs.631672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223628
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270726.1
NP_001257655.1
6
1426
NM_001270727.1
NP_001257656.1
5
1282
NM_001270728.1
NP_001257657.1
4
1262
NM_001270729.1
NP_001257658.1
6
1440
NM_001270730.1
NP_001257659.1
5
1269
NM_001270731.1
NP_001257660.1
6
1527
NM_001270732.1
NP_001257661.1
5
1370
NM_001270733.1
NP_001257662.1
2
812
NM_001270734.1
NP_001257663.1
5
1383
NM_001270735.1
NP_001257664.1
4
1226
NM_015367.3
NP_056182.2
7
1597
NR_073068.1
5
1195
NR_073069.1
5
1178
XM_011546119.1
XP_011544421.1
7
1960
XM_011546120.1
XP_011544422.1
7
1962
XM_011546121.2
XP_011544423.1
8
1945
XM_011546123.1
XP_011544425.1
6
1325
XM_017028725.1
XP_016884214.1
6
1816
XM_017028726.1
XP_016884215.1
9
2246
XM_017028727.1
XP_016884216.1
7
1395
XM_017028728.1
XP_016884217.1
7
1974
XM_017028729.1
XP_016884218.1
7
1469
XM_017028730.1
XP_016884219.1
5
1183
XM_017028731.1
XP_016884220.1
6
1803
XM_017028732.1
XP_016884221.1
4
1046
XM_017028734.1
XP_016884223.1
6
1904
GenBank mRNA
AF146568.1
-
4
1018
AF183411.1
-
1168
AF325209.1
-
6
1244
AF539453.1
-
5
1128
AK027443.1
-
1
72
AK056248.1
-
6
1283
AK291449.1
-
5
1190
AK294286.1
-
4
1089
AK301394.1
-
2
726
AK314489.1
-
7
1424
AL133029.1
-
2
699
AL831982.2
-
5
1157
BC003032.1
-
3
857
BC007658.2
-
7
1356
3343994
BC016037.1
-
1
143
CR456521.1
-
7
1500
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0