Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015425.4 | NP_056240.2 | 18 | 2997 | ||
GenBank mRNA | AK294447.1 | - | 7 | 1226 | ||
AK302458.1 | - | 18 | 2722 | |||
AK304131.1 | - | 5 | 974 | |||
AK307490.1 | - | 18 | 2717 | |||
BC073968.1 | - | 18 | 2668 | |||
BC117173.1 | - | 18 | 2650 | 40125724 | ||
BC126303.1 | - | 18 | 2650 | 8992019 | ||
BC143345.1 | - | 18 | 2650 | 9051850 | ||
U33460.1 | - | 18 | 2688 |