Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018959.3 | NP_061832.2 | 6, 7 | 673 | ||
NM_170711.2 | NP_733829.1 | 6, 7 | 673 | |||
XM_005259531.3 | XP_005259588.1 | 6, 7 | 623 | |||
XM_011527904.2 | XP_011526206.1 | 6, 7 | 1938 | |||
XM_011527905.2 | XP_011526207.1 | 6, 7 | 1946 | |||
XM_011527906.2 | XP_011526208.1 | 6, 7 | 1943 | |||
XM_011527907.2 | XP_011526209.1 | 6, 7 | 1952 | |||
XM_011527908.2 | XP_011526210.1 | 6, 7 | 1945 | |||
XM_011527909.2 | XP_011526211.1 | 6, 7 | 1945 | |||
XM_011527910.2 | XP_011526212.1 | 6, 7 | 1025 | |||
XM_017026581.1 | XP_016882070.1 | 6, 7 | 668 | |||
GenBank mRNA | AF181719.1 | - | 6, 7 | 576 | ||
AK056850.1 | - | 4, 5 | 296 | |||
BC012062.1 | - | 6, 7 | 541 | 4549444 | ||
BU557183.1 | - | 670 | ||||
KU178593.1 | - | 6, 7 | 468 |