Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDUA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000203.4 | NP_000194.2 | 5, 6 | 683 | ||
NR_110313.1 | 5, 6 | 683 | ||||
XM_011513461.2 | XP_011511763.1 | 4, 5 | 828 | |||
GenBank mRNA | AK098805.1 | - | 4, 5 | 651 | ||
AK125223.1 | - | 4, 5 | 495 | |||
AK291816.1 | - | 5, 6 | 616 | |||
BC029959.1 | - | 5, 6 | 607 | |||
M74715.1 | - | 5, 6 | 683 |