Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL2 SIRNA Assays ›
          Gene Symbol:
          IL2
          Gene Name:
          interleukin 2
          Gene Aliases:
          IL-2, TCGF, lymphokine
          Chromosome Location:
          Chr.4: 122449479 - 122456495 on Build GRCh38
          UniGene ID:
          Hs.89679
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s223932

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000586.3 NP_000577.2 3 328
          XM_017008177.1 XP_016863666.1 3 294
          GenBank mRNA A14844.1 - 3 283
          AB451325.1 - 3 273
          AB451454.1 - 3 273
          AF228636.1 - 3 285
          AY283686.1 - 3 213
          AY523040.1 - 3 213
          BC066254.1 - 3 291 6971690
          BC066255.1 - 3 291 6971691
          BC066256.1 - 3 291 6971693
          BC066257.1 - 3 291 6971694
          BC070338.1 - 3 320 30418817
          DQ231169.1 - 3 213
          DQ861285.1 - 3 275
          S77834.1 - 3 320
          S77835.1 - 3 262
          S82692.1 - 3 567
          V00564.1 - 3 320
          X01586.1 - 3 380

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          negative regulation of protein phosphorylation
          adaptive immune response
          negative regulation of B cell apoptotic process
          immune response
          cell adhesion
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          cell-cell signaling
          positive regulation of cell proliferation
          natural killer cell activation
          T cell differentiation
          positive regulation of cell growth
          positive regulation of B cell proliferation
          positive regulation of interferon-gamma production
          positive regulation of interleukin-17 production
          positive regulation of tissue remodeling
          positive regulation of activated T cell proliferation
          positive regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          response to ethanol
          positive regulation of regulatory T cell differentiation
          negative regulation of heart contraction
          positive regulation of transcription from RNA polymerase II promoter
          regulation of T cell homeostatic proliferation
          positive regulation of isotype switching to IgG isotypes
          negative regulation of lymphocyte proliferation
          negative regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of immunoglobulin secretion
          positive regulation of dendritic spine development
          extrinsic apoptotic signaling pathway in absence of ligand
          Ras guanyl-nucleotide exchange factor activity
          cytokine activity
          interleukin-2 receptor binding
          growth factor activity
          kinase activator activity
          carbohydrate binding
          kappa-type opioid receptor binding
          glycosphingolipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bfkrn:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0