Search
Search

KRT7| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005556.3 | NP_005547.3 | 4, 5 | 819 | ||
| XM_011538325.1 | XP_011536627.1 | 4, 5 | 772 | |||
| XM_017019294.1 | XP_016874783.1 | 4, 5 | 545 | |||
| GenBank mRNA | AF509887.1 | - | 4, 5 | 748 | ||
| AJ238246.1 | - | 4, 5 | 753 | |||
| AK025881.1 | - | 4, 5 | 746 | |||
| AK128505.1 | - | 4, 5 | 638 | |||
| AK128848.1 | - | 5, 6 | 674 | |||
| AK129924.1 | - | 5, 6 | 753 | |||
| AK225498.1 | - | 4, 5 | 742 | |||
| BC002700.2 | - | 4, 5 | 734 | 3610347 | ||
| BC107082.1 | - | 1, 2 | 78 | |||
| BE744321.1 | - | 396 | ||||
| X03212.1 | - | 4, 5 | 753 | |||
| Z36852.1 | - | 2, 3 | 159 | |||