Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOX4 SIRNA Assays ›
Gene Symbol:
NOX4
Gene Name:
NADPH oxidase 4
Gene Aliases:
KOX, KOX-1, RENOX
Chromosome Location:
Chr.11: 89324353 - 89589611 on Build GRCh38
UniGene ID:
Hs.371036
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224161
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NOX4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143836.2
NP_001137308.1
10, 11
1324
NM_001143837.1
NP_001137309.1
13, 14
1469
NM_001291926.1
NP_001278855.1
10, 11
1320
NM_001291927.1
NP_001278856.1
10, 11
1074
NM_001291929.1
NP_001278858.1
9, 10
987
NM_001300995.1
NP_001287924.1
10, 11
1047
NM_016931.4
NP_058627.1
10, 11
1324
XM_006718849.3
XP_006718912.1
10, 11
1252
XM_011542857.2
XP_011541159.1
10, 11
1124
XM_017017841.1
XP_016873330.1
9, 10
1171
XM_017017842.1
XP_016873331.1
6, 7
623
XM_017017843.1
XP_016873332.1
10, 11
1252
XM_017017844.1
XP_016873333.1
11, 12
1310
XM_017017845.1
XP_016873334.1
7, 8
726
GenBank mRNA
AB041035.1
-
10, 11
1244
AF254621.1
-
10, 11
1091
AF261943.1
-
10, 11
1005
AJ704725.1
-
10, 11
1001
AJ704726.1
-
10, 11
1005
AK057189.1
-
13, 14
1469
AK291830.1
-
10, 11
1087
AK298323.1
-
10, 11
1018
AK298328.1
-
10, 11
1074
AK298357.1
-
9, 10
978
AK298376.1
-
6, 7
765
AY288918.1
-
10, 11
1005
BC040105.1
-
10, 11
1077
5590269
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: BC138918, AF276957, AF218723, BC145462, BC021378.
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
cell morphogenesis
response to hypoxia
superoxide metabolic process
inflammatory response
cell aging
negative regulation of cell proliferation
gene expression
positive regulation of smooth muscle cell migration
superoxide anion generation
positive regulation of apoptotic process
positive regulation of MAP kinase activity
bone resorption
homocysteine metabolic process
positive regulation of stress fiber assembly
positive regulation of protein kinase B signaling
cardiac muscle cell differentiation
oxidation-reduction process
positive regulation of ERK1 and ERK2 cascade
cellular response to cAMP
cellular response to glucose stimulus
cellular response to gamma radiation
cellular response to transforming growth factor beta stimulus
reactive oxygen species metabolic process
positive regulation of reactive oxygen species metabolic process
positive regulation of DNA biosynthetic process
nucleotide binding
protein binding
electron carrier activity
NAD(P)H oxidase activity
superoxide-generating NADPH oxidase activity
oxygen sensor activity
heme binding
flavin adenine dinucleotide binding
modified amino acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0