Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PAX2 SIRNA Assays ›
          Gene Symbol:
          PAX2
          Gene Name:
          paired box 2
          Gene Aliases:
          FSGS7, PAPRS
          Chromosome Location:
          Chr.10: 100732940 - 100829941 on Build GRCh38
          UniGene ID:
          Hs.155644
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s224171

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PAX2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000278.4 NP_000269.3 3 788
          NM_001304569.1 NP_001291498.1 4 331
          NM_003987.4 NP_003978.3 3 788
          NM_003988.4 NP_003979.2 3 788
          NM_003989.4 NP_003980.3 3 788
          NM_003990.4 NP_003981.3 3 788
          XM_017016281.1 XP_016871770.1 4 3100
          XM_017016282.1 XP_016871771.1 4 3100
          XM_017016283.1 XP_016871772.1 4 3100
          XM_017016284.1 XP_016871773.1 4 3100
          XM_017016285.1 XP_016871774.1 4 3100
          XM_017016286.1 XP_016871775.1 4 3100
          XM_017016287.1 XP_016871776.1 4 3100
          XM_017016288.1 XP_016871777.1 4 3097
          XM_017016289.1 XP_016871778.1 4 3100
          GenBank mRNA AY153483.1 - 2 1090
          L25597.1 - 3 781
          M89470.1 - 3 781

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          urogenital system development
          branching involved in ureteric bud morphogenesis
          cell fate determination
          mesonephros development
          neural tube closure
          optic cup morphogenesis involved in camera-type eye development
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          retinal pigment epithelium development
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          axonogenesis
          mesodermal cell fate specification
          aging
          visual perception
          glial cell differentiation
          optic nerve development
          optic nerve morphogenesis
          optic nerve structural organization
          vestibulocochlear nerve formation
          response to nutrient levels
          regulation of metanephros size
          ureter maturation
          pronephric field specification
          inner ear morphogenesis
          camera-type eye development
          negative regulation of apoptotic process
          negative regulation of programmed cell death
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          protein kinase B signaling
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of cytolysis
          positive regulation of transcription from RNA polymerase II promoter
          pronephros development
          brain morphogenesis
          stem cell differentiation
          positive regulation of epithelial cell proliferation
          oxidation-reduction process
          mesenchymal to epithelial transition
          optic chiasma development
          cellular response to hydrogen peroxide
          cellular response to retinoic acid
          cellular response to glucose stimulus
          cellular response to epidermal growth factor stimulus
          metanephric mesenchyme development
          positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          metanephric mesenchymal cell differentiation
          nephric duct formation
          ureter development
          metanephric collecting duct development
          metanephric epithelium development
          metanephric distal convoluted tubule development
          metanephric nephron tubule formation
          positive regulation of metanephric glomerulus development
          negative regulation of mesenchymal cell apoptotic process involved in metanephric nephron morphogenesis
          regulation of metanephric nephron tubule epithelial cell differentiation
          reactive oxygen species metabolic process
          cochlea development
          cochlea morphogenesis
          positive regulation of branching involved in ureteric bud morphogenesis
          negative regulation of mesenchymal cell apoptotic process involved in metanephros development
          negative regulation of apoptotic process involved in metanephric collecting duct development
          negative regulation of apoptotic process involved in metanephric nephron tubule development
          negative regulation of reactive oxygen species metabolic process
          positive regulation of metanephric DCT cell differentiation
          positive regulation of optic nerve formation
          core promoter proximal region sequence-specific DNA binding
          DNA binding
          protein binding
          transcription factor binding
          superoxide-generating NADPH oxidase activity
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0