Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004578.3 | NP_004569.2 | 2 | 330 | ||
NR_073545.1 | 2 | 330 | ||||
GenBank mRNA | AF498934.1 | - | 2 | 52 | ||
AK223009.1 | - | 2 | 95 | |||
AK313807.1 | - | 2 | 303 | |||
AY585832.1 | - | 2 | 259 | |||
BC002438.1 | - | 2 | 123 | 3346455 | ||
BC004309.1 | - | 2 | 260 | 3627110 | ||
BI828974.1 | - | 2 | 214 | |||
DA712952.1 | - | 2 | 330 | |||
HY142709.1 | - | 2 | 310 | |||
KC407675.1 | - | 2 | 52 | |||
M28211.1 | - | 2 | 107 | |||
X13902.1 | - | 2 | 163 |