siRNA Details
Gene Symbol: ROCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005406.2 | NP_005397.1 | 6, 7 | 1608 | ||
GenBank mRNA | AK057038.1 | - | 6, 7 | 1609 | ||
AK307388.1 | - | 6, 7 | 1423 | |||
U43195.1 | - | 6, 7 | 667 |

More Information
Other Species Targeted

Panther Classification:
