siRNA Details
Gene Symbol: SOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003107.2 | NP_003098.1 | 1 | 2033 | ||
GenBank mRNA | BC072668.1 | - | 2 | 1853 | 6584346 | |
X70683.1 | - | 1 | 1589 |

More Information
Other Species Targeted

Panther Classification:
