Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224843
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
4, 5
579
NM_001007471.2
NP_001007472.2
4, 5
927
NM_020952.4
NP_066003.3
4, 5
579
NM_024971.5
NP_079247.5
4, 5
579
NM_206944.3
NP_996827.3
4, 5
579
NM_206945.3
NP_996828.3
4, 5
579
NM_206946.3
NP_996829.3
4, 5
579
NM_206947.3
NP_996830.3
4, 5
579
NM_206948.2
NP_996831.1
4, 5
579
XM_011519035.2
XP_011517337.1
4, 5
804
XM_011519036.2
XP_011517338.1
4, 5
3729
XM_011519037.2
XP_011517339.1
4, 5
3715
XM_011519038.2
XP_011517340.1
4, 5
3712
XM_011519039.2
XP_011517341.1
4, 5
3713
XM_011519040.2
XP_011517342.1
4, 5
810
XM_011519041.2
XP_011517343.1
4, 5
3712
XM_011519042.2
XP_011517344.1
4, 5
810
XM_011519043.2
XP_011517345.1
4, 5
843
XM_011519044.2
XP_011517346.1
4, 5
3713
XM_011519045.2
XP_011517347.1
4, 5
804
XM_011519046.2
XP_011517348.1
4, 5
2507
XM_011519047.2
XP_011517349.1
4, 5
2477
XM_017015141.1
XP_016870630.1
4, 5
804
XM_017015142.1
XP_016870631.1
4, 5
804
XM_017015143.1
XP_016870632.1
4, 5
804
XM_017015144.1
XP_016870633.1
4, 5
570
XM_017015145.1
XP_016870634.1
4, 5
1248
XM_017015146.1
XP_016870635.1
4, 5
646
XM_017015147.1
XP_016870636.1
4, 5
2681
XM_017015148.1
XP_016870637.1
4, 5
804
XM_017015149.1
XP_016870638.1
4, 5
804
XM_017015150.1
XP_016870639.1
4, 5
804
XM_017015151.1
XP_016870640.1
4, 5
822
XM_017015152.1
XP_016870641.1
4, 5
2482
XM_017015153.1
XP_016870642.1
4, 5
814
XM_017015154.1
XP_016870643.1
4, 5
950
XM_017015155.1
XP_016870644.1
4, 5
2479
XM_017015157.1
XP_016870646.1
4, 5
2477
XM_017015158.1
XP_016870647.1
4, 5
2475
XM_017015159.1
XP_016870648.1
4, 5
570
XM_017015160.1
XP_016870649.1
4, 5
660
XM_017015161.1
XP_016870650.1
4, 5
804
GenBank mRNA
AB099661.1
-
1, 2
224
AB099662.1
-
1, 2
224
AB099663.1
-
1, 2
224
AB099664.1
-
1, 2
224
AB099665.1
-
1, 2
224
AF325212.1
-
4, 5
550
AF536748.1
-
1, 2
224
AF536749.1
-
1, 2
224
AF536750.1
-
1, 2
224
AF536751.1
-
1, 2
224
AF536752.1
-
1, 2
224
AF536753.1
-
1, 2
224
AJ505025.1
-
4, 5
927
AJ505026.1
-
4, 5
927
AK021788.1
-
4, 5
570
AK225732.1
-
4, 5
570
AK308682.1
-
4, 5
755
AL136545.1
-
4, 5
760
BC022454.2
-
3, 4
509
BC067733.1
-
4, 5
541
BC094699.1
-
4, 5
573
BC121821.1
-
2, 3
425
BC134414.1
-
2, 3
428
BC142972.1
-
4, 5
596
CD104154.1
-
4, 5
575
KF987075.1
-
4, 5
635
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0