Search
Search

SELO| Gene Symbol: SELO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031454.1 | NP_113642.1 | 5, 6 | 1415 | ||
| GenBank mRNA | AY324823.1 | - | 5, 6 | 1415 | ||
| BC001099.2 | - | 5, 6 | 905 | |||
| BC020510.1 | - | 4, 5 | 726 | |||
| BC110866.1 | - | 5, 6 | 1385 | 6041782 | ||
| CR456464.1 | - | 5, 6 | 931 | |||