Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014913.3 | NP_055728.1 | 4 | 876 | ||
XM_005266656.4 | XP_005266713.1 | 4 | 730 | |||
XM_011525883.2 | XP_011524185.1 | 4 | 590 | |||
GenBank mRNA | AB020670.1 | - | 4 | 606 | ||
AK292566.1 | - | 4 | 645 | |||
BC071589.1 | - | 4 | 876 | 30333923 |