Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MADD SIRNA Assays ›
          Gene Symbol:
          MADD
          Gene Name:
          MAP kinase activating death domain
          Gene Aliases:
          DENN, IG20, RAB3GEP
          Chromosome Location:
          Chr.11: 47269376 - 47330031 on Build GRCh38
          UniGene ID:
          Hs.82548
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s225001

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MADD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135943.1 NP_001129415.1 19 3355
          NM_001135944.1 NP_001129416.1 19 3331
          NM_003682.3 NP_003673.3 20 3520
          NM_130470.2 NP_569826.2 19 3626
          NM_130471.2 NP_569827.2 20 3391
          NM_130472.2 NP_569828.2 19 3331
          NM_130473.2 NP_569829.2 20 3520
          NM_130474.2 NP_569830.2 19 3331
          NM_130475.2 NP_569831.1 20 3544
          NM_130476.2 NP_569832.2 19 3484
          XM_005253189.2 XP_005253246.1 20 3544
          XM_005253196.2 XP_005253253.1 20 3544
          XM_005253199.2 XP_005253256.1 19 3484
          XM_005253200.2 XP_005253257.1 20 3544
          XM_005253201.2 XP_005253258.1 20 3415
          XM_005253203.2 XP_005253260.1 19 3355
          XM_005253204.2 XP_005253261.1 19 3355
          XM_005253205.1 XP_005253262.1 19 3355
          XM_017018478.1 XP_016873967.1 20 3544
          XM_017018479.1 XP_016873968.1 20 3544
          XM_017018480.1 XP_016873969.1 20 3544
          XM_017018481.1 XP_016873970.1 19 3484
          XM_017018482.1 XP_016873971.1 20 3544
          XM_017018483.1 XP_016873972.1 20 3415
          XM_017018484.1 XP_016873973.1 19 3484
          XM_017018485.1 XP_016873974.1 19 3445
          XM_017018486.1 XP_016873975.1 20 3415
          XM_017018487.1 XP_016873976.1 19 3355
          XM_017018488.1 XP_016873977.1 20 3544
          XM_017018489.1 XP_016873978.1 20 3505
          XM_017018490.1 XP_016873979.1 20 3531
          XM_017018491.1 XP_016873980.1 20 3500
          XM_017018492.1 XP_016873981.1 20 3544
          XM_017018493.1 XP_016873982.1 19 3471
          XM_017018494.1 XP_016873983.1 19 3484
          XM_017018495.1 XP_016873984.1 19 3484
          XM_017018496.1 XP_016873985.1 19 3440
          XM_017018497.1 XP_016873986.1 19 3445
          XM_017018498.1 XP_016873987.1 20 3415
          XM_017018499.1 XP_016873988.1 20 3384
          XM_017018500.1 XP_016873989.1 19 3484
          XM_017018501.1 XP_016873990.1 20 3371
          XM_017018502.1 XP_016873991.1 20 3384
          XM_017018503.1 XP_016873992.1 20 3371
          XM_017018504.1 XP_016873993.1 20 3384
          XM_017018505.1 XP_016873994.1 20 3371
          XM_017018506.1 XP_016873995.1 20 3376
          XM_017018507.1 XP_016873996.1 19 3342
          XM_017018508.1 XP_016873997.1 19 3355
          XM_017018509.1 XP_016873998.1 19 3311
          XM_017018510.1 XP_016873999.1 19 3355
          GenBank mRNA AB002356.1 - 20 3516
          AF440100.1 - 20 3494
          AF440101.1 - 20 3494
          AF440102.1 - 20 3365
          AF440103.1 - 19 3305
          AF440434.1 - 19 3305
          AK292442.1 - 19 3295
          BC040484.1 - 19 3339 5271798
          U44953.1 - 19 3448
          U77352.1 - 19 3594

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053585, U72995.

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          cell surface receptor signaling pathway
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of Rab protein signal transduction
          regulation of apoptotic process
          positive regulation of GTPase activity
          regulation of cell cycle
          execution phase of apoptosis
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of extrinsic apoptotic signaling pathway
          death receptor binding
          protein binding
          Rab guanyl-nucleotide exchange factor activity
          protein kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0