Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM2 SIRNA Assays ›
Gene Symbol:
R3HDM2
Gene Name:
R3H domain containing 2
Gene Aliases:
CAG6, PR01365
Chromosome Location:
Chr.12: 57253764 - 57430999 on Build GRCh38
UniGene ID:
Hs.443673
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22531
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014925.3
NP_055740.3
18
2167
XM_005268725.3
XP_005268782.1
20
2517
XM_011538031.1
XP_011536333.1
21
2405
XM_011538033.1
XP_011536335.1
22
2459
XM_011538035.1
XP_011536337.1
21
2363
XM_011538036.1
XP_011536338.1
21
2357
XM_011538037.1
XP_011536339.1
20
2339
XM_011538038.1
XP_011536340.1
20
2309
XM_011538039.1
XP_011536341.1
20
2303
XM_011538040.1
XP_011536342.1
21
2357
XM_011538042.2
XP_011536344.1
23
2555
XM_017018998.1
XP_016874487.1
20
2381
XM_017018999.1
XP_016874488.1
23
2596
XM_017019000.1
XP_016874489.1
21
2363
XM_017019001.1
XP_016874490.1
22
2669
XM_017019002.1
XP_016874491.1
20
2333
XM_017019003.1
XP_016874492.1
20
2309
XM_017019004.1
XP_016874493.1
20
2303
XM_017019005.1
XP_016874494.1
22
2514
XM_017019006.1
XP_016874495.1
21
2407
XM_017019007.1
XP_016874496.1
21
2442
XM_017019008.1
XP_016874497.1
19
2207
XM_017019009.1
XP_016874498.1
20
2261
XM_017019010.1
XP_016874499.1
20
2335
XM_017019011.1
XP_016874500.1
23
2558
XM_017019012.1
XP_016874501.1
19
2243
XM_017019013.1
XP_016874502.1
20
2377
XM_017019014.1
XP_016874503.1
20
2412
XM_017019015.1
XP_016874504.1
20
2554
XM_017019016.1
XP_016874505.1
21
2573
XM_017019017.1
XP_016874506.1
20
2335
XM_017019018.1
XP_016874507.1
20
2301
XM_017019019.1
XP_016874508.1
21
2397
XM_017019020.1
XP_016874509.1
18
2147
XM_017019021.1
XP_016874510.1
20
2371
XM_017019022.1
XP_016874511.1
19
2281
XM_017019023.1
XP_016874512.1
19
2292
XM_017019024.1
XP_016874513.1
19
2281
XM_017019025.1
XP_016874514.1
19
2246
XM_017019026.1
XP_016874515.1
20
2286
XM_017019027.1
XP_016874516.1
19
2417
XM_017019028.1
XP_016874517.1
18
2179
XM_017019029.1
XP_016874518.1
18
2265
XM_017019030.1
XP_016874519.1
11
1546
XM_017019031.1
XP_016874520.1
14
1830
XM_017019032.1
XP_016874521.1
10
1398
XM_017019033.1
XP_016874522.1
12
1549
GenBank mRNA
AB023219.1
-
20
2524
AK293394.1
-
10
1419
AX775761.1
-
20
2524
AX775763.1
-
20
2566
BC041857.1
-
10
1979
5270511
BC104995.1
-
8
1174
8143998
BC112226.1
-
8
1174
8327694
BC143605.1
-
9
1276
9052118
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0