Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42EP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006779.3 | NP_006770.1 | 2 | 1815 | ||
XM_017017094.1 | XP_016872583.1 | 2 | 1926 | |||
GenBank mRNA | AK098292.1 | - | 2 | 1929 | ||
BC022337.1 | - | 1 | 1410 | 4690444 |