Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NLGN1 SIRNA Assays ›
          Gene Symbol:
          NLGN1
          Gene Name:
          neuroligin 1
          Gene Aliases:
          NL1
          Chromosome Location:
          Chr.3: 173395899 - 174291287 on Build GRCh38
          UniGene ID:
          Hs.478289
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22547

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NLGN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014932.3 NP_055747.1 4 1049
          XM_005247234.2 XP_005247291.1 3 1224
          XM_005247235.3 XP_005247292.1 3 1420
          XM_005247236.2 XP_005247293.1 3 1224
          XM_005247237.3 XP_005247294.1 2 1718
          XM_006713540.3 XP_006713603.1 4 1480
          XM_011512551.2 XP_011510853.1 5 1331
          XM_011512552.2 XP_011510854.1 4 1767
          XM_017005888.1 XP_016861377.1 4 1628
          XM_017005889.1 XP_016861378.1 4 1628
          XM_017005890.1 XP_016861379.1 4 1284
          XM_017005891.1 XP_016861380.1 4 1480
          XM_017005892.1 XP_016861381.1 4 1782
          XM_017005893.1 XP_016861382.1 5 1894
          XM_017005894.1 XP_016861383.1 4 3412
          XM_017005895.1 XP_016861384.1 4 7585
          XM_017005896.1 XP_016861385.1 4 1066
          XM_017005897.1 XP_016861386.1 4 1628
          XM_017005898.1 XP_016861387.1 3 1568
          XM_017005899.1 XP_016861388.1 3 1707
          XM_017005900.1 XP_016861389.1 4 1213
          XM_017005901.1 XP_016861390.1 3 1269
          XM_017005902.1 XP_016861391.1 4 1271
          XM_017005903.1 XP_016861392.1 3 1420
          XM_017005904.1 XP_016861393.1 3 1707
          GenBank mRNA AB028993.1 - 3 1289
          AK307813.1 - 5 1175
          BC032555.1 - 4 1054 5558890
          GQ489206.1 - 4 740

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053868, XM_008760890, U22952.
          This siRNA also targets the following transcripts in Mouse: NM_001163387, NM_138666, BC005523, BC098461, AK122433.

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of respiratory gaseous exchange by neurological system process
          protein targeting
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          neuron cell-cell adhesion
          nervous system development
          synapse assembly
          positive regulation of circadian sleep/wake cycle, wakefulness
          synaptic vesicle targeting
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          neuronal signal transduction
          neuron projection development
          positive regulation of synaptic transmission, GABAergic
          protein localization to synapse
          establishment of protein localization
          regulation of neuron differentiation
          rhythmic process
          cytoskeletal matrix organization at active zone
          protein homooligomerization
          protein heterotetramerization
          positive regulation of filopodium assembly
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          long-term synaptic potentiation
          positive regulation of dendritic spine development
          negative regulation of dendritic spine morphogenesis
          terminal button organization
          synaptic vesicle clustering
          postsynaptic membrane assembly
          presynaptic membrane assembly
          AMPA glutamate receptor clustering
          NMDA glutamate receptor clustering
          neurexin clustering involved in presynaptic membrane assembly
          postsynaptic density protein 95 clustering
          receptor localization to synapse
          positive regulation of ruffle assembly
          positive regulation of synaptic vesicle endocytosis
          positive regulation of protein localization to synapse
          positive regulation of intracellular signal transduction
          positive regulation of synaptic vesicle exocytosis
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          positive regulation of synaptic vesicle clustering
          receptor activity
          PDZ domain binding
          neurexin family protein binding
          protein dimerization activity
          cell adhesion molecule binding
          carboxylic ester hydrolase activity
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0