Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317962.1 | NP_001304891.1 | 5 | 473 | ||
NM_001317963.1 | NP_001304892.1 | 5 | 645 | |||
NM_007062.2 | NP_008993.1 | 5 | 617 | |||
GenBank mRNA | AK223162.1 | - | 5 | 541 | ||
AK290681.1 | - | 5 | 458 | |||
AK296252.1 | - | 5 | 474 | |||
AK297961.1 | - | 4 | 302 | |||
BC000067.2 | - | 5 | 500 | |||
BC001652.2 | - | 5 | 536 | 2924215 | ||
BC010921.1 | - | 5 | 579 | |||
BC032127.2 | - | 5 | 539 | |||
BC040135.1 | - | 5 | 515 | |||
BC046144.1 | - | 5 | 531 | |||
L07758.1 | - | 5 | 536 |